3-Chlorobenzoyl chloride 99+%
Leverantör: Thermo Scientific
Fara
Synonyms: m-Chlorobenzoyl chloride, 3-Chlorobenzoylchloride
108740050 108740500 108741000 108745000
ACRO108740050EA 455 SEK
ACRO108740050 ACRO108740500 ACRO108741000 ACRO108745000
3-Chlorobenzoyl chloride 99+%
3-Chlorobenzoyl chloride
Formel: C₇H₄Cl₂O
MW: 175,01 g/mol
Kokpunkt: 223 °C (1013 hPa)
Smältpunkt: 44 °C
Densitet: 1,36 g/cm³
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000671
CAS nummer: 618-46-2
EINECS: 210-552-6
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5680 to 1.5700 (20°C, 589 nm)
GC ≤0.03 % Total other poly-halogenated acid halides
Titration RCOCl ≥99.0 %
Titration RCOOH ≤1.0 % (as Chlorobenzoic acid)