3,4-Dichlorophenylboronic acid ≥97%
Leverantör: Thermo Scientific
Varning
B24292.03 B24292.06 B24292.14
B24292.03EA 850 SEK
B24292.03 B24292.06 B24292.14
3,4-Dichlorophenylboronic acid ≥97%
3,4-Dichlorophenylboronic acid
Purity: 97% CAS No.: 151169-75-4 Molecular Formula: C6H5BCl2O2 Formula Weight: 190.82 Melting Point: 270-274° Beilstein Reference: 7369790
Formel: C₆H₅BCl₂O₂
MW: 190,82 g/mol
Smältpunkt: 270…274 °C
Storage Temperature: Omgivande
MDL Number: MFCD01074646
CAS nummer: 151169-75-4