2-Bromobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Bromobenzene-1-sulphonyl chloride
A13645.14 A13645.06
A13645.14EA 8275 SEK
A13645.14 A13645.06
2-Bromobenzenesulphonyl chloride ≥98%
2-Bromobenzenesulphonyl chloride
Purity: 98% CAS No.: 2905-25-1 Molecular Formula: C6H4BrClO2S Formula Weight: 255.52 Melting Point: 48-52° Boiling Point: 127-128°/2.4mm UN No.: 3261 MDL No.: MFCD00051971
Formel: C₆H₄BrClO₂S
MW: 255,52 g/mol
Kokpunkt: 127…128 °C (2,4 mmHg)
Smältpunkt: 48…52 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051971
CAS nummer: 2905-25-1
UN: 3261
ADR: 8,II