2,5-Dibromobenzenesulphonyl chloride ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,5-Dibromobenzene-1-sulphonyl chloride
L19888.03 L19888.06
L19888.03EA 1650 SEK
L19888.03 L19888.06
2,5-Dibromobenzenesulphonyl chloride ≥99%
2,5-Dibromobenzenesulphonyl chloride
Purity: 99% CAS No.: 23886-64-8 Molecular Formula: C6H3Br2ClO2S Formula Weight: 334.41 Melting Point: 69-72° Beilstein Reference: 2694054 UN No.: 3261 MDL No.: MFCD00060685
Formel: C₆H₃Br₂ClO₂S
MW: 334,42 g/mol
Smältpunkt: 69…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00060685
CAS nummer: 23886-64-8
UN: 3261
ADR: 8,II