2,4-Dibromobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,4-Dibromobenzene-1-sulphonyl chloride
L19887.03 L19887.06
L19887.03EA 1725 SEK
L19887.03 L19887.06
2,4-Dibromobenzenesulphonyl chloride ≥98%
2,4-Dibromobenzenesulphonyl chloride
Purity: 98% CAS No.: 72256-95-2 Molecular Formula: C6H3Br2ClO2S Formula Weight: 334.41 Melting Point: 82-86° Boiling Point: 128°/0.2mm UN No.: 3261 MDL No.: MFCD03093759
Formel: C₆H₃Br₂ClO₂S
MW: 334,42 g/mol
Kokpunkt: 128 °C (0,2 mmHg)
Smältpunkt: 82…86 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093759
CAS nummer: 72256-95-2
UN: 3261
ADR: 8,II