2,6-Dibromo-4-nitrophenol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3-Dibromo-2-hydroxy-5-nitrobenzene
A12916.14 A12916.22 A12916.30
A12916.14EA 1175 SEK
A12916.14 A12916.22 A12916.30
2,6-Dibromo-4-nitrophenol ≥98%
2,6-Dibromo-4-nitrophenol
Purity: 98% CAS No.: 99-28-5 Molecular Formula: C6H3Br2NO3 Formula Weight: 296.90 Melting Point: ca 145° dec. Beilstein Reference: 1245050
Formel: C₆H₃Br₂NO₃
Smältpunkt: ∼ 145 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007334
CAS nummer: 99-28-5
EINECS: 202-744-3