4-Bromophenyl isothiocyanate
Leverantör: Thermo Scientific
Synonyms:
1-Bromo-4-isothiocyanatobenzene, 4-Bromophenylisothiocyanate
SB01925EA
SB01925EE
MAYBSB01925EAEA
0
SEK
MAYBSB01925EA
MAYBSB01925EE
4-Bromophenyl isothiocyanate
4-Bromophenyl isothiocyanate
Formel:
C₇H₄BrNS MW: 214.09 g/mol Kokpunkt: 142…144 °C (14 mmHg) Smältpunkt: 60 Storage Temperature: Omgivande |
MDL Number:
MFCD00004808 CAS nummer: 1985-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...