2-Bromo-5-fluorophenyl isothiocyanate

Leverantör: Thermo Scientific

SB01713DA SB01713EA
MAYBSB01713DAEA 0 SEK
MAYBSB01713DA MAYBSB01713EA
2-Bromo-5-fluorophenyl isothiocyanate
2-Bromo-5-fluorophenyl isothiocyanate
Formel: C₇H₃BrFNS
MW: 232.08 g/mol
MDL Number: MFCD00276607
CAS nummer: 1027513-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR