1-Bromo-2,4,6-trifluorobenzen ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Bromo-1,3,5-trifluorobenzene
A19920.06 A19920.14
A19920.06EA 1500 SEK
A19920.06 A19920.14
1-Bromo-2,4,6-trifluorobenzen ≥98%
1-Bromo-2,4,6-trifluorobenzen
Purity: 98+% CAS No.: 2367-76-2 Molecular Formula: C6H2BrF3 Formula Weight: 210.98 Flash Point: 58°(136°F) Refractive Index: 1.4850 Melting Point: 3-4° Boiling Point: 140-141° Beilstein Reference: 2502668 UN No.: 1993 MDL No.: MFCD00000334
Formel: C₆H₂BrF₃
MW: 210,98 g/mol
Kokpunkt: 140…141 °C
Smältpunkt: 3…4 °C
Densitet: 1,790
Flampunkt: 58 °C (136 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000334
CAS nummer: 2367-76-2
EINECS: 219-125-9
UN: 1993
ADR: 3,III