3-Bromo-2,6-dimethoxybenzoic acid
Leverantör: Thermo Scientific
BTB07593DA
BTB07593EA
MAYBBTB07593DAEA
0
SEK
MAYBBTB07593DA
MAYBBTB07593EA
3-Bromo-2,6-dimethoxybenzoic acid
3-Bromo-2,6-dimethoxybenzoic acid
Formel:
C₉H₉BrO₄ |
MDL Number:
MFCD00052933 CAS nummer: 73219-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...