Methyl-4-bromobenzoate ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: Methyl 4-bromobenzoate, 4-Bromobenzoic acid methyl ester
A15517.06 A15517.14 A15517.22
A15517.06EA 1150 SEK
A15517.06 A15517.14 A15517.22
Methyl-4-bromobenzoate ≥98%
Methyl-4-bromobenzoate
Purity: 98+% CAS No.: 619-42-1 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 77-81° Beilstein Reference: 2045132 Merck Reference: 14,1408 MDL No.: MFCD00013531
Formel: C₈H₇BrO₂
Kokpunkt: 246 °C (1013 hPa)
Smältpunkt: 77…81 °C
Densitet: 1,69
Storage Temperature: Omgivande
MDL Number: MFCD00013531
CAS nummer: 619-42-1
EINECS: 210-596-6
Merck Index: 14,1408