Methyl-3-bromobenzoate ≥98%
Leverantör: Thermo Scientific
Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester
A16174.09 A16174.18 A16174.30
A16174.09EA 1150 SEK
A16174.09 A16174.18 A16174.30
Methyl-3-bromobenzoate ≥98%
Methyl-3-bromobenzoate
Purity: 98+% CAS No.: 618-89-3 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Flash Point: 110°(230°F) Refractive Index: 1.5575 Melting Point: 30-33° Boiling Point: 139-140°/35mm Beilstein Reference: 1940810 MDL No.: MFCD00017777
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
Kokpunkt: 139…140 °C (35 mmHg)
Smältpunkt: 30…33 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00017777
CAS nummer: 618-89-3
EINECS: 210-569-9