Methyl-3-bromobenzoate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester
333650050 333650250
ACRO333650050EA 605 SEK
ACRO333650050 ACRO333650250
Methyl-3-bromobenzoate 98%
Methyl-3-bromobenzoate
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
Kokpunkt: 139…140 °C (35 mmHg)
Smältpunkt: 31 °C
MDL Number: MFCD00017777
CAS nummer: 618-89-3
EINECS: 210-569-9
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting crystalline solid or liquid
Infrared spectrum Conforms
HPLC ≥97.5 %