4-Bromobenzoyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: p-Bromobenzoyl chloride
L03794.09 L03794.18
L03794.09EA 980 SEK
L03794.09 L03794.18
4-Bromobenzoyl chloride ≥98%
4-Bromobenzoyl chloride
Purity: 98+% CAS No.: 586-75-4 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 110°(230°F) Melting Point: 38-42° Boiling Point: 246° Beilstein Reference: 636641 UN No.: 3261 MDL No.: MFCD00000683
Formel: C₇H₄BrClO
Kokpunkt: 246 °C
Smältpunkt: 38…42 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000683
CAS nummer: 586-75-4
EINECS: 209-580-1
UN: 3261
ADR: 8,II