3-Bromobenzoyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride
A15256.09 A15256.18 A15256.30
A15256.09EA 825 SEK
A15256.09 A15256.18 A15256.30
3-Bromobenzoyl chloride ≥98%
3-Bromobenzoyl chloride
Purity: 98% CAS No.: 1711-09-7 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 107°(224°F) Refractive Index: 1.5950 Boiling Point: 247-251° Beilstein Reference: 1635505 UN No.: 3265 MDL No.: MFCD00000669
Formel: C₇H₄BrClO
Kokpunkt: 247…251 °C
Densitet: 1,666
Flampunkt: 107 °C (224 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000669
CAS nummer: 1711-09-7
EINECS: 216-978-9
UN: 3265
ADR: 8,II