5-Bromo-2-klorofluorobenzen ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Bromo-1-chloro-2-fluorobenzene
A15745.14 A15745.22
A15745.14EA 2075 SEK
A15745.14 A15745.22
5-Bromo-2-klorofluorobenzen ≥98%
5-Bromo-2-klorofluorobenzen
Purity: 98% CAS No.: 60811-18-9 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Refractive Index: 1.5550 Boiling Point: 62-64°/9mm MDL No.: MFCD00042519
Formel: C₆H₃BrClF
MW: 209,44 g/mol
Kokpunkt: 62…64 °C (9 mmHg)
Densitet: 1,710
Storage Temperature: Omgivande
MDL Number: MFCD00042519
CAS nummer: 60811-18-9