3-Bromo-4-chlorobenzoic acid ≥97%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L19244.03
L19244.06
L19244.03EA
1450
SEK
L19244.03
L19244.06
3-Bromo-4-chlorobenzoic acid ≥97%
3-Bromo-4-chlorobenzoic acid
Purity: 97%
CAS No.: 42860-10-6
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Melting Point: 218-222°
Beilstein Reference: 2438796
Formel:
C₇H₄BrClO₂ Smältpunkt: 218…222 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00079706 CAS nummer: 42860-10-6 |