4-Bromophenylboronic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid
107280050 107280250
ACRO107280050EA 1550 SEK
ACRO107280050 ACRO107280250
4-Bromophenylboronic acid 98%
4-Bromophenylboronic acid
Formel: C₆H₆BBrO₂
Smältpunkt: 287…292 °C
Densitet: 1,67 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002104
CAS nummer: 5467-74-3
EINECS: 226-779-9
Order Now
Specifikation testresultat
Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %