2-Bromophenylboronic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: (2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid
379990050 379990250 379991000
ACRO379990050EA 1750 SEK
ACRO379990050 ACRO379990250 ACRO379991000
2-Bromophenylboronic acid 97%
2-Bromophenylboronic acid
Formel: C₆H₆BBrO₂
MW: 200,83 g/mol
Smältpunkt: 109…114 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01114672
CAS nummer: 244205-40-1
Order Now
Specifikation testresultat
Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %