4-Hydroxyphenylboronic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
369420050 369420010
ACRO369420050EA 3525 SEK
ACRO369420050 ACRO369420010
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Formel: C₆H₇BO₃
MW: 137,93 g/mol
Smältpunkt: 285 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD01074628
CAS nummer: 71597-85-8
Order Now
Specifikation testresultat
Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %