3,4-Dimethoxyphenylboronic acid ≥98%
Leverantör: Thermo Scientific
Varning
B24240.03 B24240.06 B24240.14
B24240.03EA 1225 SEK
B24240.03 B24240.06 B24240.14
3,4-Dimethoxyphenylboronic acid ≥98%
3,4-Dimethoxyphenylboronic acid
Purity: 98% CAS No.: 122775-35-3 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: ca 240° Beilstein Reference: 5334877
Formel: C₈H₁₁BO₄
MW: 181,98 g/mol
Smältpunkt: ∼ 240 °C
Storage Temperature: Omgivande
MDL Number: MFCD01074574
CAS nummer: 122775-35-3