5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
B23661.06 B23661.14
B23661.06EA 2150 SEK
B23661.06 B23661.14
5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97%
5-Bromo-2,2-difluoro-1,3-benzodioxole
Purity: 97% CAS No.: 33070-32-5 Molecular Formula: C7H3BrF2O2 Formula Weight: 237.00 Refractive Index: 1.4967 Boiling Point: 78-79°/20mm MDL No.: MFCD00236212
Formel: C₇H₃BrF₂O₂
MW: 237 g/mol
Kokpunkt: 78…79 °C (20 mmHg)
Densitet: 1,740
Storage Temperature: Omgivande
MDL Number: MFCD00236212
CAS nummer: 33070-32-5