1,4-naftokinon, (max. 5% H₂O) 97+%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
A10958.0E A10958.22 A10958.36
A10958.0EEA 6975 SEK
A10958.0E A10958.22 A10958.36
1,4-naftokinon, (max. 5% H₂O) 97+%
1,4-naftokinon
Purity: 97+% CAS No.: 130-15-4 Molecular Formula: C10H6O2 Formula Weight: 158.15 Flash Point: 142°(288°F) Odour: Irritating Beilstein Reference: 878524 Merck Reference: 14,6395 UN No.: 2811 MDL No.: MFCD00001676
Formel: C₁₀H₆O₂
Kokpunkt: 298 °C (1013 hPa)
Smältpunkt: 120…124 °C
Densitet: 1,420
Flampunkt: 142 °C (288 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001676
CAS nummer: 130-15-4
EINECS: 204-977-6
UN: 2811
ADR: 6.1,I
Merck Index: 14,6395