2-Indolinone 97%, ren
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
129670250 129671000 129670050
ACRO129670250EA 3025 SEK
ACRO129670250 ACRO129671000 ACRO129670050
2-Indolinone 97%, ren
2-Indolinone
Formel: C₈H₇NO
MW: 133,15 g/mol
Kokpunkt: 227 °C (73 mmHg)
Smältpunkt: 123…128 °C
Densitet: 1,32 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005711
CAS nummer: 59-48-3
EINECS: 200-429-5
Order Now
Specifikation testresultat
Appearance Beige to yellow to orange to pink-brown Powder
Infrared spectrum Conforms
Melting point 123 to 128 °C
HPLC ≥96.0 %