Benzimidazole ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole
A12763.0B A12763.18 A12763.30
A12763.0BEA 6150 SEK
A12763.0B A12763.18 A12763.30
Benzimidazole ≥99%
Benzimidazole
Purity: 99% CAS No.: 51-17-2 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 170-174° Boiling Point: 360° Merck Reference: 14,1081 MDL No.: MFCD00005585
Formel: C₇H₆N₂
Kokpunkt: 360 °C
Smältpunkt: 170…174 °C
Densitet: 1,23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005585
CAS nummer: 51-17-2
EINECS: 200-081-4
Merck Index: 14,1081