Pempidine ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2,2,6,6-Pentamethylpiperidine
H27729.06 H27729.03
H27729.06EA 1975 SEK
H27729.06 H27729.03
Pempidine ≥97%
Pempidine
Purity: 97% CAS No.: 79-55-0 Molecular Formula: C10H21N Formula Weight: 155.28 Flash Point: 50°(122°F) Odour: Amine-like Boiling Point: 187-188° Beilstein Reference: 103806 Merck Reference: 14,7080 UN No.: 1992 MDL No.: MFCD00006493
Formel: C₁₀H₂₁N
MW: 155,28 g/mol
Kokpunkt: 187…188 °C
Smältpunkt: 11 °C
Densitet: 0,858
Flampunkt: 50 °C (122 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006493
CAS nummer: 79-55-0
EINECS: 201-211-2
UN: 1992
ADR: 3,III
Merck Index: 14,7080