2,2,6,6-Tetramethylpiperidine ≥98%
Leverantör: Thermo Scientific
Fara
A18712.06 A18712.14 A18712.22
A18712.06EA 910 SEK
A18712.06 A18712.14 A18712.22
2,2,6,6-Tetramethylpiperidine ≥98%
2,2,6,6-Tetramethylpiperidine
Purity: 98+% CAS No.: 768-66-1 Molecular Formula: C9H19N Formula Weight: 141.26 Flash Point: 34°(93°F) Refractive Index: 1.4440 Odour: Amine-like Melting Point: -59° Boiling Point: 152-154° Beilstein Reference: 103296 UN No.: 2733 MDL No.: MFCD00005985
Formel: C₉H₁₉N
MW: 141,26 g/mol
Kokpunkt: 152…154 °C
Smältpunkt: –59 °C
Densitet: 0,832
Flampunkt: 34 °C (93 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005985
CAS nummer: 768-66-1
EINECS: 212-199-3
ADR: 3,III