N-Metyl-2-pyrrolidon (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
364381000 364380010 364380025 364385000
364381000.EA 675 SEK
364381000. 364380010. 364380025. 364385000.
N-Metyl-2-pyrrolidon (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
N-Metyl-2-pyrrolidon (NMP)
Formel: C₅H₉NO
Kokpunkt: 202 °C (760 mmHg)
Smältpunkt: –24 °C
Densitet: 1,03 g/cm³
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003193
CAS nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.5 %
Color scale ≤50 APHA