9-Bromoanthracene ≥99.0% (by GC and HPLC)
Leverantör: TCI EUROPE
Varning
B2871.0005 B2871-100G B2871-25G B2871-5G
TCIAB2871.0005EA 0 SEK
TCIAB2871.0005 TCIAB2871-100G TCIAB2871-25G TCIAB2871-5G
9-Bromoanthracene ≥99.0% (by GC and HPLC)
9-Bromoanthracene
Formel: C₁₄H₉Br
MW: 257,13 g/mol
Kokpunkt: 359 °C (1013 hPa)
Smältpunkt: 103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001243
CAS nummer: 1564-64-3
EINECS: 216-359-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99756e5a5ca473172198600985a7e05e46b2162&vid=08b7566b9c14dfa4810528ef1ea0dbf08bc6b209&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 99.0 %
Purity (HPLC) min. 99.0 area%
Melting point 103.0 to 107.0 deg-C
Solubility in Toluene almost transparency