4,4-Dimethyl-2-pentyne ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: t-Butylmethylacetylene, tert-Butyl methyl acetylene, 4,4-Dimethylpent-2-yne
L10210.03 L10210.06
L10210.03EA 1325 SEK
L10210.03 L10210.06
4,4-Dimethyl-2-pentyne ≥97%
4,4-Dimethyl-2-pentyne
Purity: 97+% CAS No.: 999-78-0 Molecular Formula: C7H12 Formula Weight: 96.17 Flash Point: -10°(14°F) Refractive Index: 1.4085 Odour: Petrol-like Boiling Point: 82° Beilstein Reference: 1733540 UN No.: 3295 MDL No.: MFCD00041613
Formel: C₇H₁₂
MW: 96,17 g/mol
Kokpunkt: 82 °C
Smältpunkt: –82,4 °C
Densitet: 0,718
Flampunkt: –10 °C (14 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00041613
CAS nummer: 999-78-0
ADR: 3,II