4-Pentyn-1-ol 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Pentyn-5-ol
330430050 330430250
ACRO330430050EA 1550 SEK
ACRO330430050 ACRO330430250
4-Pentyn-1-ol 95%
4-Pentyn-1-ol
Formel: C₅H₈O
MW: 84,12 g/mol
Kokpunkt: 154…155 °C
Densitet: 0,91 g/cm³
Flampunkt: 61 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002974
CAS nummer: 5390-04-5
EINECS: 226-383-6
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4470 (20°C, 589 nm)
GC ≥94 %