4-Pentyn-2-ol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Pentyn-4-ol
L02752.06 L02752.14
L02752.06EA 1100 SEK
L02752.06 L02752.14
4-Pentyn-2-ol ≥97%
4-Pentyn-2-ol
Purity: 97% CAS No.: 2117-11-5 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 37°(98°F) Refractive Index: 1.4380 Boiling Point: 126-127° Beilstein Reference: 1734245 UN No.: 1987 MDL No.: MFCD00004555
Formel: C₅H₈O
MW: 84,12 g/mol
Kokpunkt: 126…127 °C
Densitet: 0,892
Flampunkt: 37 °C (98 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004555
CAS nummer: 2117-11-5
EINECS: 218-321-1
UN: 1987
ADR: 3,III