10-Undecin-1-ol 96%
Leverantör: Thermo Scientific
Synonyms: 10-Undecyn-1-ol, Undec-10-yn-1-ol, 1-Undecyn-11-ol
L11807.06 L11807.03
L11807.06EA 3475 SEK
L11807.06 L11807.03
10-Undecin-1-ol 96%
10-Undecin-1-ol
Purity: 96% CAS No.: 2774-84-7 Molecular Formula: HC≡C(CH2)9OH Formula Weight: 168.28 Refractive Index: 1.4565 Boiling Point: 110-111°/4mm MDL No.: MFCD00041675
Formel: C₁₁H₂₀O
MW: 168,28 g/mol
Kokpunkt: 110…111 °C (4 mmHg)
Densitet: 0,873
Storage Temperature: Omgivande
MDL Number: MFCD00041675
CAS nummer: 2774-84-7