3-Pentyn-1-ol 99%
Leverantör: Thermo Fisher Scientific
Varning
197330050 197330250 197331000
ACRO197330050EA 1600 SEK
ACRO197330050 ACRO197330250 ACRO197331000
3-Pentyn-1-ol 99%
3-Pentyn-1-ol
Formel: C₅H₈O
Kokpunkt: 154…157 °C (760 mmHg)
Densitet: 0,91 g/cm³
Flampunkt: 54 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002956
CAS nummer: 10229-10-4
EINECS: 233-550-7
UN: 1987
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥98.5 %