3-Octyn-1-ol 96%
Leverantör: Thermo Fisher Scientific
L05414.06 L05414.14
L05414.06EA 1375 SEK
L05414.06 L05414.14
3-Octyn-1-ol 96%
3-Octyn-1-ol
Purity: 96% CAS No.: 14916-80-4 Molecular Formula: CH3(CH2)3C≡C(CH2)2OH Formula Weight: 126.20 Flash Point: 110°(230°F) Refractive Index: 1.4540 Boiling Point: 86-88°/9mm Beilstein Reference: 1739950 MDL No.: MFCD00039552
Formel: C₈H₁₄O
Kokpunkt: 86…88 °C (9 mmHg)
Densitet: 0,883
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039552
CAS nummer: 14916-80-4
EINECS: 238-986-1