2-Octyn-1-ol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
|
L05596.09
L05596.18
L05596.09EA
1850
SEK
L05596.09
L05596.18
2-Octyn-1-ol ≥98%
2-Octyn-1-ol
Purity: 98%
CAS No.: 20739-58-6
Molecular Formula: CH3(CH2)4C≡CCH2OH
Formula Weight: 126.20
Flash Point: 91°(195°F)
Refractive Index: 1.4560
Boiling Point: 76-78°/2mm
Beilstein Reference: 1744120
MDL No.: MFCD00039542
Formel:
C₈H₁₄O Kokpunkt: 76…78 °C (2 mmHg) Densitet: 0,880 Flampunkt: 91 °C (195 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00039542 CAS nummer: 20739-58-6 |