2-Hexyn-1-ol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Hex-2-yn-1-ol
A13339.14 A13339.06 A13339.09
A13339.14EA 5800 SEK
A13339.14 A13339.06 A13339.09
2-Hexyn-1-ol ≥97%
2-Hexyn-1-ol
Purity: 97% CAS No.: 764-60-3 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 68°(154°F) Refractive Index: 1.4540 Boiling Point: 167° Beilstein Reference: 1736274 MDL No.: MFCD00039540
Formel: C₆H₁₀O
MW: 98,14 g/mol
Kokpunkt: 167 °C
Densitet: 0,897
Flampunkt: 68 °C (154 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039540
CAS nummer: 764-60-3
EINECS: 212-128-6