2-Butynoic acid ethyl ester ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl 2-butynoate
B25370.03 B25370.06 B25370.14
B25370.03EA 715 SEK
B25370.03 B25370.06 B25370.14
2-Butynoic acid ethyl ester ≥98%
2-Butynoic acid ethyl ester
Purity: 98% CAS No.: 4341-76-8 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 53°(127°F) Refractive Index: 1.4380 Odour: Characteristic Boiling Point: 163° Beilstein Reference: 1744948 UN No.: 3272 MDL No.: MFCD00015182
Formel: C₆H₈O₂
Kokpunkt: 163 °C
Densitet: 0,967
Flampunkt: 53 °C (127 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00015182
CAS nummer: 4341-76-8
EINECS: 224-395-6
UN: 3272
ADR: 3,III