5-Chloro-1-pentyne 98%
Leverantör: Thermo Fisher Scientific
Fara
223420050 223420250
ACRO223420050EA 1150 SEK
ACRO223420050 ACRO223420250
5-Chloro-1-pentyne 98%
5-Chloro-1-pentyne
Formel: C₅H₇Cl
MW: 102.56 g/mol
Kokpunkt: 67…69 °C (145 mmHg)
Densitet: 0.96 g/cm³
Flampunkt: 15 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001014
CAS nummer: 14267-92-6
EINECS: 238-159-5
UN: 1993
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥97.5 %