1,4-Dichloro-2-butyne 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1,4-Dichlorobut-2-yne
113280250 113281000
ACRO113280250EA 2850 SEK
ACRO113280250 ACRO113281000
1,4-Dichloro-2-butyne 99%
1,4-Dichloro-2-butyne
Formel: C₄H₄Cl₂
MW: 122,98 g/mol
Kokpunkt: 165…168 °C (760 mmHg)
Smältpunkt: -34,5 °C
Densitet: 1,25 g/cm³
Flampunkt: 160 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000981
CAS nummer: 821-10-3
EINECS: 212-474-8
UN: 2810
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5060 (20°C, 589 nm)
GC ≥98.5 %