Kanelalkohol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
148520050 148521000 148522500 148525000
ACRO148520050EA 435 SEK
ACRO148520050 ACRO148521000 ACRO148522500 ACRO148525000
Kanelalkohol ≥98%
Kanelalkohol
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokpunkt: 256 °C (1013 hPa)
Smältpunkt: 31…35 °C
Densitet: 1,04 g/cm³
Flampunkt: 126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002921
CAS nummer: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325
Order Now
Specifikation testresultat
Appearance White to light yellow Fused crystalline solid
Infrared spectrum Conforms
Melting point 31 to 35 °C
GC ≥97.5 %