Caffeic acid predominantly trans ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
A15950.09 A15950.18
A15950.09EA 1500 SEK
A15950.09 A15950.18
Caffeic acid predominantly trans ≥98%
Caffeic acid
Purity: 99% CAS No.: 331-39-5 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: ca 202° dec. Beilstein Reference: 2210883 Merck Reference: 14,1635 MDL No.: MFCD00004392
Formel: C₉H₈O₄
MW: 180,16 g/mol
Kokpunkt: 366 °C (1013 hPa)
Smältpunkt: ∼ 202 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00004392
CAS nummer: 331-39-5
EINECS: 206-361-2
Merck Index: 14,1635