3-Methyl-3-buten-1-ol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methylbut-3-en-1-ol, 2-Methyl-1-buten-4-ol
156631000 156635000
ACRO156631000EA 865 SEK
ACRO156631000 ACRO156635000
3-Methyl-3-buten-1-ol 97%
3-Methyl-3-buten-1-ol
Formel: C₅H₁₀O
MW: 86,13 g/mol
Kokpunkt: 132 °C (1013 hPa)
Smältpunkt: –82 °C
Densitet: 0,85 g/cm³
Flampunkt: 36 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002933
CAS nummer: 763-32-6
EINECS: 212-110-8
UN: 1987
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4327 to 1.4347 (20°C, 589 nm)
GC ≥96.0 %
Titration with HClO4 ≥98.5 %
Insoluble matter ≤0.0005 %