2-Methyl-3-buten-2-ol 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Hydroxy-3-methyl-1-butene
159670010 159672500
ACRO159670010EA 1600 SEK
ACRO159670010 ACRO159672500
2-Methyl-3-buten-2-ol 97%
2-Methyl-3-buten-2-ol
Formel: C₅H₁₀O
MW: 86,13 g/mol
Kokpunkt: 99 °C (1013 hPa)
Smältpunkt: –43 °C
Densitet: 0,82 g/cm³
Flampunkt: 13 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004470
CAS nummer: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥96.0 %
Elemental Analysis 8.35 to 9.42 % (N)