cis-3-Octen-1-ol 95%
Leverantör: Thermo Scientific
Varning
Synonyms: cis-Oct-3-en-1-ol
A19014.09 A19014.18
A19014.09EA 1025 SEK
A19014.09 A19014.18
cis-3-Octen-1-ol 95%
cis-3-Octen-1-ol
Purity: 95% CAS No.: 20125-84-2 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 82°(179°F) Refractive Index: 1.4470 Boiling Point: 88-89°/15mm Beilstein Reference: 1720075 MDL No.: MFCD00014062
Formel: C₈H₁₆O
MW: 128,21 g/mol
Kokpunkt: 88…89 °C (15 mmHg)
Densitet: 0,850
Flampunkt: 82 °C (179 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014062
CAS nummer: 20125-84-2
EINECS: 243-533-6