Ethyl-3,4,5-trihydroxybenzoate ≥98%
Leverantör: Thermo Scientific
Synonyms: Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
A13733.09 A13733.18
A13733.09EA 1050 SEK
A13733.09 A13733.18
Ethyl-3,4,5-trihydroxybenzoate ≥98%
Ethyl-3,4,5-trihydroxybenzoate
Purity: 98% CAS No.: 831-61-8 Molecular Formula: C9H10O5 Formula Weight: 198.18 Melting Point: 148-152° Beilstein Reference: 2116014
Formel: C₉H₁₀O₅
MW: 198,18 g/mol
Smältpunkt: 148…152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016430
CAS nummer: 831-61-8
EINECS: 212-608-5