1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: Diethylene glycol divinyl ether, 1,1'-[Oxybis(ethyleneoxy)]diethene, 1,1'-(Oxybis(ethyleneoxy))diethylene, 1,1'-(Oxybis(ethyleneoxy))diethene, Diethyleneglycoldivinylether, (2-(2-Ethenyloxyethoxy)ethoxy)ethene
Stabilization: Stabilised with potassium hydroxide 0,1%
L12501.09 L12501.18
L12501.09EA 1025 SEK
L12501.09 L12501.18
1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98% stabilised
1,1'-[Oxybis(ethyleneoxy)]diethylene
Stabilized 0.1% potassium hydroxide
Purity: 98% CAS No.: 764-99-8 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 83°(181°F) Refractive Index: 1.4470 Melting Point: -21° Boiling Point: 198-199° Beilstein Reference: 1752316 MDL No.: MFCD00054610
Formel: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MW: 158,2 g/mol
Kokpunkt: 198…199 °C
Smältpunkt: –21 °C
Densitet: 0,968
Flampunkt: 83 °C (181 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00054610
CAS nummer: 764-99-8
EINECS: 212-133-3