Allyl isothiocyanate ≥94% stabilised
Leverantör: Thermo Fisher Scientific
Fara
L02901.06 L02901.18 L02901.30
L02901.06EA 730 SEK
L02901.06 L02901.18 L02901.30
Allyl isothiocyanate ≥94% stabilised
Allyl isothiocyanate
Stabilized
Purity: 94% CAS No.: 57-06-7 Molecular Formula: CH2=CHCH2NCS Formula Weight: 99.16 Storage Temperature: 2°C to 8°C Flash Point: 46°(114°F) Refractive Index: 1.5300 Odour: Pungent Melting Point: -80° Boiling Point: 149-151° Beilstein Reference: 773748 Merck Reference: 14,295 UN No.: 1545 MDL No.: MFCD00004822
Formel: C₄H₅NS
MW: 99.16 g/mol
Kokpunkt: 149…151 °C
Smältpunkt: –80 °C
Densitet: 1.013
Flampunkt: 46 °C (114 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00004822
CAS nummer: 57-06-7
EINECS: 200-309-2
UN: 1545
ADR: 6.1 II
Merck Index: 13,00292