4-Pentenoic acid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Allylacetic acid, Pent-4-enoic acid
269020250 269020050 269021000
ACRO269020250EA 2200 SEK
ACRO269020250 ACRO269020050 ACRO269021000
4-Pentenoic acid 99%
4-Pentenoic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 83…84 °C (12 mmHg)
Smältpunkt: –22,5 °C
Densitet: 0,975 g/cm³
Flampunkt: 89 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004408
CAS nummer: 591-80-0
EINECS: 209-732-7
UN: 3265
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4273 to 1.4303 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25°C) 0.971 to 0.979