Dimetylmaleat 96%
Leverantör: Thermo Scientific
Varning
Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester
218460010 218462500 218460051
ACRO218460010EA 2500 SEK
ACRO218460010 ACRO218462500 ACRO218460051
Dimetylmaleat 96%
Dimetylmaleat
Formel: C₆H₈O₄
MW: 144,13 g/mol
Kokpunkt: 204…205 °C (760 mmHg)
Smältpunkt: –17,5 °C
Densitet: 1,15 g/cm³
Flampunkt: 95 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008459
CAS nummer: 624-48-6
EINECS: 210-848-5
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥95 %
Color scale ≤20 APHA